Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200055

Supelco

Monoclonal Anti-Glutathione-S-Transferase (GST) antibody produced in rat

clone GST-R 6G9, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GST-R 6G9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~27 kDa

should not react with

human, rat, bovine, rabbit

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 0.05-0.1 μg/mL using The detection limit fGST GST recombinant protein is ~1.0 μg/lane under non-reducing and reducing conditions.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GSTP1(2950)

Specificity

Monoclonal Anti-Glutathione S-Transferase (GST)is specific for GST from Schistosoma japonicum, and does not recognize GST from rabbit liver, porcine liver, bovine liver, rat liver or human placenta when tested.

Immunogen

GST fusion protein

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at -20 °C. For continuous use, the product may be stored at 2-8 °C for up to one month. For extended storage, freeze at at -20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Orosa et al.
Nature communications, 9(1), 5185-5185 (2018-12-07)
Detection of conserved microbial patterns by host cell surface pattern recognition receptors (PRRs) activates innate immunity. The FLAGELLIN-SENSITIVE 2 (FLS2) receptor perceives bacterial flagellin and recruits another PRR, BAK1 and the cytoplasmic-kinase BIK1 to form an active co-receptor complex that
Irina Kufareva et al.
Scientific reports, 9(1), 15893-15893 (2019-11-07)
CK2 is a constitutively active protein kinase overexpressed in numerous malignancies. Interaction between CK2α and CK2β subunits is essential for substrate selectivity. The CK2α/CK2β interface has been previously targeted by peptides to achieve functional effects; however, no small molecules modulators
Benoît Bestgen et al.
Journal of medicinal chemistry, 62(4), 1817-1836 (2019-01-29)
Protein CK2 has gained much interest as an anticancer drug target in the past decade. We had previously described the identification of a new allosteric site on the catalytic α-subunit, along with first small molecule ligands based on the 4-(4-phenylthiazol-2-ylamino)benzoic
Edward J Strini et al.
Frontiers in plant science, 13, 857745-857745 (2022-04-22)
The final shape and size of plant organs are determined by a network of genes that modulate cell proliferation and expansion. Among those, SCI1 (Stigma/style Cell-cycle Inhibitor 1) functions by inhibiting cell proliferation during pistil development. Alterations in SCI1 expression
Moumita Srivastava et al.
Current biology : CB, 30(8), 1410-1423 (2020-02-29)
Brassinosteroids (BRs) play crucial roles in plant development, but little is known of mechanisms that integrate environmental cues into BR signaling. Conjugation to the small ubiquitin-like modifier (SUMO) is emerging as an important mechanism to transduce environmental cues into cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service