Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200051

Sigma-Aldrich

Anti-JUB (C-terminal) antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Ajuba

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~57 kDa

species reactivity

human, mouse

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using P19 cells
indirect immunofluorescence: 5-10 μg/mL using HeLa cells and 10-20 μg/mL using P19 cells
western blot: 0.3-0.6 μg/mL using HEK-293T cell lysates expressing human JUB

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JUB(84962)

General description

JUB, also known as Ajuba, is encoded by the gene mapped to human chromosome 14q11.2. JUB is an adaptor/scaffolding protein and is a member of the zyxin family of cytosolic LIM proteins. The protein is characterized with three related tandem LIM domains at the C terminus associated with unique and distinctive N-terminal, or Pre LIM domains.

Specificity

Anti-JUB (C-terminal) specifically recognizes human and mouse JUB.

Application

Anti-JUB (C-terminal) antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

JUB mediates various cellular events. It plays a role in cadherin-mediated cell-cell adhesion and in cell migration. JUB is recruited to cadherin complexes through interaction with α catenin, and also influences the localization of p130Cas at focal adhesions and Rac1 activity during cell migration. Altered expression of the gene has been observed in human papillomavirus (HPV) virus.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The LIM protein, Limd1, regulates AP-1 activation through an interaction with Traf6 to influence osteoclast development
Feng Y, et al.
The Journal of Biological Chemistry, 282(1), 39-48 (2007)
The LIM protein Ajuba influences p130Cas localization and Rac1 activity during cell migration
Pratt SJ, et al.
The Journal of Cell Biology, 168(5), 813-824 (2005)
Genomic insights into head and neck cancer
Beck TN and Golemis EA
Cancer, 1(1), 1-1 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service