Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200050

Sigma-Aldrich

Anti-SRGAP3 antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Wave-associated rac GTPase-activating protein , Anti-ARHGAP14, Anti-MEGAP, Anti-Mental disorder-associated gap protein, Anti-SLIT-ROBO GAP 3, Anti-SLIT-ROBO Rho GTPase-activating protein 3 (SRGAP3), Anti-WRP

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$422.00

$422.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$422.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$422.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~124 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3 μg/mL using mouse brain extracts (S1 fraction) and HEK-293T cell lysates overexpressing SRGAP3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OXSR1(9901)

General description

Anti-SRGAP3 is produced in rabbit using as the immunogen a synthetic peptide corresponding to a fragment of human SRGAP3 (GeneID: 9901), conjugated to KLH. SRGAP3 (Slit-ROBO Rho GTPase-activating protein 3, also known as MEGAP, is a GTPase activating protein, that belongs to the Slit-ROBO GAPs (SRGAPs) family. It is predominantly expressed in the brain during embryonic development and in the adult brain, specifically in neurons of the hippocampus and cortex. SRGAP3 exists as two isoforms SRGAP3A and SRGAP3B.

Application

Anti-SRGAP3 antibody produced in rabbit has been used in immunofluorescence and immunoblotting.

Biochem/physiol Actions

SRGAP3 (Slit-ROBO Rho GTPase-activating protein 3) negatively regulates cell migration, by inhibiting the activity of Rac1 and Cdc42. SRGAP3/MEGAP has been associated with severe mental retardation in several patients. It plays pivotal roles in learning and memory. SRGAP3/WRP interacts directly to WAVE-1 through its Src homology domain and specifically inhibits Rac function in vivo.Induced expression of SRGAP3 inhibits Rac1 and Cdc42 activity and impedes cell migration by impairing actin and microtubule dynamics and formation of focal complexes.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MEGAP impedes cell migration via regulating actin and microtubule dynamics and focal complex formation
Yang Y, et al.
Experimental Cell Research, 312(12), 2379-2393 (2006)
Signal transduction in neuronal migration: roles of GTPase activating proteins and the small GTPase Cdc42 in the Slit-Robo pathway
Wong K, et al.
Cell, 107(2), 209-221 (2001)
The WRP component of the WAVE-1 complex attenuates Rac-mediated signalling
Soderling SH, et al.
Nature Cell Biology, 4(12), 970-970 (2002)
SrGAP3 interacts with lamellipodin at the cell membrane and regulates Rac-dependent cellular protrusions
Endris V, et al.
Journal of Cell Science, 124(23), 3941-3955 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service