Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200038

Sigma-Aldrich

Anti-SORBS2 (N-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARGBP2, Anti-Arg binding protein 2, Anti-Arg/Abl-interacting protein 2, Anti-Sorbin and SH3 domain containing 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~85 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

indirect immunofluorescence: 20-40 μg/mL using HeLa cells
western blot: 0.2-0.4 μg/mL using HEK-293T cell lysates expressing human SORBS2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORBS2(8470)

General description

SORBS2 (also known as ArgBP2), is an Arg/Abl (proto-oncogene 1, non-receptor tyrosine kinase)-binding protein, that contains an N-terminal sorbin homology (SoHo) domain which interacts with lipid raft proteins, three C-terminal SH3 domains, a Ser/Thr-rich domain, and several potential Abl phosphorylation sites. SORBS2 associates with and is a substrate of Arg and v-Abl. The human SORBS2 gene encodes multiple transcripts that yield different isoforms. SORBS2 is ubiquitously expressed in human tissues and predominant in heart and epithelial cells. SORBS2 is also found in stress fibers and in the nucleus. In cardiac muscle cells, SORBS2 is in the Z-disks of sarcomeres.

Application

Anti-SORBS2 (N-terminal) antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Sorbin and SH3 domain containing 2 (SORBS2) might function as an adapter protein to assemble signaling complexes in stress fibers and is a potential link between Abl kinases and the actin cytoskeleton.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

nArgBP2, a novel neural member of ponsin/ArgBP2/vinexin family that interacts with synapse-associated protein 90/postsynaptic density-95-associated protein (SAPAP)
Kawabe H, et al.
Test, 274(43), 30914-30918 (1999)
The Abl/Arg substrate ArgBP2/nArgBP2 coordinates the function of multiple regulatory mechanisms converging on the actin cytoskeleton
Cestra G, et al.
Proceedings of the National Academy of Sciences of the USA, 102(5), 1731-1736 (2005)
Vinexin, CAP/ponsin, ArgBP2: a novel adaptor protein family regulating cytoskeletal organization and signal transduction
Kioka N, et al.
Cell Structure and Function, 27(1), 1-7 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service