Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200021

Sigma-Aldrich

Anti-OS9 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERLEC2, Anti-Endoplasmic reticulum associated protein, Anti-Osteosarcoma amplified 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~97/88 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of rat NRK cells
western blot: 4-8 μg/mL using whole extracts of human HEK-293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OS9(10956)
mouse ... Os9(216440)
rat ... Os9(362891)

General description

OS9 endoplasmic reticulum lectin is encoded by the gene mapped to human chromosome 12q13.3-q14.1. Alternative splicing of the OS-9 gene yields two proteins, namely OS-9.1 and OS-9.2. These proteins are ubiquitously expressed in human tissues.

Specificity

Anti-OS9 recognizes human, rat, and mouse OS9 isoforms 1 and 2.

Application

Anti-OS9 antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

OS9 endoplasmic reticulum lectin is an endoplasmic reticulum (ER)-resident lectin involved in ER quality control. OS9 is upregulated in response to ER stress through activation of the Ire1/Xbp1 pathway. OS9 associates both with components of the ERAD machinery and ERAD substrates. OS9 binds to misfolding proteins and prevents their secretion from the ER. It then directs them to the suppressor/enhancer of Lin-12-like (Sel1L) / hydroxymethylglutaryl reductase degradation protein 1 (Hrd1) dislocation and ubiquitylation complex in the ER membrane. Two OS9 spliced variants, OS-9.1 and OS-9.2 expressions are amplified in tumors.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mammalian OS-9 is upregulated in response to endoplasmic reticulum stress and facilitates ubiquitination of misfolded glycoproteins
Alcock F and Swanton E
Journal of Molecular Biology, 385(4), 1032-1042 (2009)
Neue pathogenetische Konzepte und fr
Schupp JC, et al.
Pneumologie (Stuttgart, Germany), 70(04), 231-240 (2016)
Enterovirus 71 protease 2Apro and 3Cpro differentially inhibit the cellular endoplasmic reticulum-associated degradation (ERAD) pathway via distinct mechanisms, and enterovirus 71 hijacks ERAD component p97 to promote its replication
Wang T, et al.
PLoS Pathogens, 13(10), e1006674-e1006674 (2017)
A Dual Task for the Xbp1-responsive OS-9 Variants in the Mammalian Endoplasmic Reticulum INHIBITING SECRETION OF MISFOLDED PROTEIN CONFORMERS AND ENHANCING THEIR DISPOSAL
Bernasconi R, et al.
The Journal of biological chemistry, 283(24), 16446-16454 (2008)
Chromosomal and genetic imbalances in Chinese patients with rhabdomyosarcoma detected by high-resolution array comparative genomic hybridization
Liu C, et al.
International Journal of Clinical and Experimental Pathology, 7(2), 690-690 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service