Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200016

Sigma-Aldrich

Anti-SLC9A1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-APNH, Anti-NHE1, Anti-Na-Li counter transporter, Anti-Porter, Sodium-hydrogen ion, amiloride-sensitive, Anti-Sodium/hydrogen exchanger 1, Anti-Solute carrier family 9, Isoform A1, Anti-Solute carrier family 9, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 2.5-5.0 μg/mL using whole extract of human platelets

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC9A1(6548)

General description

The solute carrier family 9 member A1 (SLC9A1) gene is mapped to human chromosome 1p36.11. The encoded protein is widely expressed in many tissues and is localized to the plasma membrane. SLC9A1 is the only isoform found in mammalian cardiomyocytes. This gene encodes a 815 amino acid protein containing 12 transmembrane spanning domain and a cytoplasmic domain with many regulatory sites.

Application

Anti-SLC9A1 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Solute carrier family 9 member A1 (SLC9A1) mediates the electroneutral exchange of Na+-H+ in response to intracellular acidosis. It drives extracellular Na+ uptake in exchange for intracellular H+. SLC9A1 maintains the pH and cell volume regulation of vertebrate cells. It is specifically inhibited by the diuretic drug amiloride and activated by a variety of signals including growth factors, mitogens, neurotransmitters, tumor promoters and cell shrinkage.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physiological functions and regulation of the Na+/H+ exchanger [NHE1] in renal tubule epithelial cells
Valles PG, et al.
Kidney & Blood Pressure Research, 40(5), 452-466 (2015)
Na+-H+ exchanger-1 (NHE1) regulation in kidney proximal tubule
Parker MD, et al.
Cellular and Molecular Life Sciences, 72(11), 2061-2074 (2015)
Sodium-Hydrogen Exchanger Isoform-1 Inhibition: A Promising Pharmacological Intervention for Resuscitation from Cardiac Arrest
Gazmuri R, et al.
Molecules (Basel), 24(9), 1765-1765 (2019)
CHP1-mediated NHE1 biosynthetic maturation is required for Purkinje cell axon homeostasis
Liu Ye, et al.
The Journal of Neuroscience, 33(31), 12656-12669 (2013)
Naoko Fujii et al.
The Journal of biological chemistry, 291(47), 24787-24799 (2016-10-14)
Hypotonic stress decreased claudin-1 and -2 expression levels in renal tubular epithelial HK-2 and Madin-Darby canine kidney cells. Here, we examined the regulatory mechanism involved in this decrease. The hypotonicity-induced decrease in claudin expression was inhibited by the following: SB202190

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service