Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200014

Sigma-Aldrich

Anti-TRIP6 antibody, Mouse monoclonal

clone TRP(N)-12, purified from hybridoma cell culture

Synonym(s):

Anti-MGC10556, Anti-MGC10558, Anti-MGC29959, Anti-MGC3837, Anti-MGC4423, Anti-OIP1, Anti-Thyroid hormone receptor interactor 6, Anti-ZRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

TRP(N)-12, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human, bovine

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using whole extract of human HeLa cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIP6(7205)

General description

Monoclonal Anti-TRIP6 (mouse IgG1 isotype) is derived from the hybridoma TRP(N)-12 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Thyroid hormone receptor interactor 6 (TRIP6) is a member of the zyxin family. The TRIP6 gene encodes a protein with three Lin11, Isl-1, Mec-3 (LIM) zinc-binding domains at its carboxy- terminus, a proline-rich region and nuclear export signals at its aminoterminus. TRIP6 transcripts are strongly detected in highly vascularized tissues such as lungs, placenta, heart, and kidney. This protein localizes to focal adhesion sites and along actin stress fibers. The TRIP6 gene is mapped on the human chromosome at 7q22.1.

Specificity

Monoclonal Anti-TRIP6 recognizes human and bovine TRIP6.

Application

Anti-TRIP6 antibody, Mouse monoclonal may be used in immunoblotting.

Biochem/physiol Actions

Thyroid hormone receptor interactor 6 (TRIP6) is recruited to the plasma membrane in a lysophosphatidic acid (LPA)-dependent manner and it also regulates LPA-induced cell migration. This protein is postulated to serve as a docking site for the assembly of multimeric protein complexes involved in regulating cytoskeleton assembly and cell motility.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Xu et al.
The Journal of biological chemistry, 279(11), 10459-10468 (2003-12-23)
Lysophosphatidic acid (LPA) induces actin rearrangement, focal adhesion assembly, and cell migration through the activation of small G protein Rho and its downstream effectors. These diverse cellular responses are mediated by its associated G protein-coupled receptors. However, the mechanisms and
Yuan Wang et al.
Biochimica et biophysica acta, 1593(2-3), 115-120 (2003-02-13)
Zyxin and paxillin are the prototypes of two related subfamilies of LIM domain proteins that are localized primarily at focal adhesion plaques. However, recent work has shown that zyxin/paxillin family proteins also shuttle through the nucleus. These proteins may enter
Jie Huang et al.
Blood, 120(24), 4873-4881 (2012-09-20)
We conducted a genome-wide association study to identify novel associations between genetic variants and circulating plasminogen activator inhibitor-1 (PAI-1) concentration, and examined functional implications of variants and genes that were discovered. A discovery meta-analysis was performed in 19 599 subjects
Jinseong Yi et al.
The Journal of biological chemistry, 277(11), 9580-9589 (2002-01-10)
Integrin binding to extracellular matrix proteins induces formation of signaling complexes at focal adhesions. Zyxin co-localizes with integrins at sites of cell-substratum adhesion and is postulated to serve as a docking site for the assembly of multimeric protein complexes involved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service