Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200013

Sigma-Aldrich

Anti-Kindlin-3 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FERMT3, Anti-Fermitin family homolog 3, Anti-KIND3, Anti-MIG-2, Anti-MIG2B, Anti-UNC-112 related protein 2, Anti-UNC112C, Anti-URP2, Anti-URP2SF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 5-10 μg/mL using whole extracts of human TF1 cells

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FERMT3(83706)

General description

Kindlin-3 is a member of the kindlin family of focal adhesion proteins. The kindlin family consists of three members in vertebrates, kindlin-1, kindlin-2 and kindlin-3 that share high sequence homology and a common structure containing a C-terminal 4.1R, ezrin, radixin, moesin (FERM) domain and a pleckstrin-homology domain. Kindlin-1 and kindlin-2 are widely expressed, kindlin-3 is preferentially expressed in hematopoietic cells, mainly in megakaryocytes and platelets.

Application

Anti-Kindlin-3 antibody produced in rabbit has been used in western blotting and immunoblotting.

Biochem/physiol Actions

Kindlin-3 is required for integrin-mediated leukocyte adhesion to endothelial cells. Mutations in kindlin-3 cause leukocyte adhesion deficiency-III. Kindlin-1, kindlin-2 and kindlin-3 are essential for the regulation of integrin activation by binding the tails of integrin-β subunits. Loss of kindlins results in severe defects in integrin signaling, cell-ECM adhesion and cytoskeletal organization. Deficiency of kindlin-1 results in kindler syndrome, kindlin-2 results in early embryonic lethality due to cardiac developmental problems, and kindlin-3 results in postnatal lethality.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kindling the flame of integrin activation and function with kindlins
Plow EF, et al.
Current Opinion in Hematology, 16(5), 323-323 (2009)
Kindlins: essential regulators of integrin signalling and cell-matrix adhesion
Larjava H, et al.
EMBO Reports, 9(12), 1203-1208 (2008)
Peripheral blood mononuclear cell proteome changes in patients with myelodysplastic syndrome
Pecankova K, et al.
BioMed Research International, 2015(3), 325-325 (2015)
Integrin associated proteins differentially regulate neutrophil polarity and directed migration in 2D and 3D
Yamahashi Y, et al.
Biomedical Microdevices, 17(5), 100-100 (2015)
The Kindlins: subcellular localization and expression during murine development
Ussar S, et al.
Experimental Cell Research, 312(16), 3142-3151 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service