Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200012

Sigma-Aldrich

Anti-TIM4 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-T-cell immunoglobulin and mucin domain-containing protein 4, Anti-T-cell membrane protein 4, Anti-TIMD4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa
antigen ~85 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of HEK-293T cells overexpressing human TIM4

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMD4(91937)

General description

T cell immunoglobulin and mucin domains containing protein 4 (TIM4, TIMD4) belong to the TIM family of proteins that are type I cell surface glycoproteins. It consists of an immunoglobulin variable (IgV) domain with six cysteines, a heavily glycosylated mucin-like domain, a transmembrane domain and a cytoplasmic domain. All TIMs contain a tyrosine phosphorylation motif in the cytoplasmic tail except for TIM4. It is expressed primarily on mature antigen-presenting cells (APCs) and is the natural ligand for TIM1.
The TIM4 gene is mapped to human chromosome 5q33.3. The encoded protein is exclusively expressed in antigen-presenting cells.

Specificity

Anti-TIM4 (N-terminal) recognizes human TIM4.

Application

Anti-TIM4 (N-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

T cell immunoglobulin and mucin domains containing protein 4 (TIM4, TIMD4) is a phosphatidylserine receptor that mediates phagocytosis of apoptotic cells. It is involved in the regulation of T helper cells (Th1 and Th2) immune responses. TIM4 binds apoptotic cells by recognizing phosphatidylserine through its immunoglobulin domain. TIM1/TIM4 interaction regulates T cell activation and proliferation.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TIM-1 and TIM-4 glycoproteins bind phosphatidylserine and mediate uptake of apoptotic cells
Kobayashi N, et al.
Immunity, 27(6), 927-940 (2007)
Eksavang Khounphinith et al.
International journal of medical sciences, 16(6), 864-871 (2019-07-25)
Background: The T-cell immunoglobulin and mucin domain 4 gene (TIMD4) rs6882076 single nucleotide polymorphism (SNP) has been associated with serum total cholesterol, low-density lipoprotein cholesterol and triglycerides (TG) levels, but the results are inconsistent. Moreover, little is known about such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service