Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200009

Sigma-Aldrich

Anti-Leupaxin antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LDPL, Anti-LPXN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~43 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using extracts of Raji cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LPXN(9404)
mouse ... Lpxn(107321)
rat ... Lpxn(293783)

General description

Leupaxin is a LIM domain phosphotyrosine protein, encoded by the gene mapped to human chromosome 11q12.1. It is a member of paxillin protein family and is preferentially expressed in macrophages, lymphoid tissues and hematopoietic cells. Leupaxin contains two types of protein interaction domains, LIM and leucine−aspartate (LD) motifs that share homology with paxillin. LIM domains have been shown to mediate localization to focal contacts. LD motifs have been implicated in focal adhesion kinase (FAK) and vinculin binding, resulting in the localization of FAK to focal adhesions. The C-terminal LIM domains in leupaxin share 70% amino acid identity with paxillin. The N-terminal region of leupaxin contains three short stretches of 13 amino acids that share 70−90% similarity with paxillin LD motifs.

Specificity

Anti-Leupaxin specifically recognizes human leupaxin.

application

Anti-Leupaxin antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Leupaxin associates with a second FAK family member, PYK2 and regulates cell type-specific signaling complex in leukocytes. It also acts as a substrate for a tyrosine kinase in lymphoid cells and thus, may function in and be regulated by tyrosine kinase activity. It can form a complex with PYK2 and PTP-PEST in several cell types including monocytes and osteoclasts and in prostate cancer cells. In osteoclasts, leupaxin has been suggested to be a critical nucleating component of the osteoclast podosomal signaling complex in the adhesion zone. It is expressed in prostate cancer and is thought to play a role in adhesion and invasion of prostate carcinoma cells.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leupaxin promotes bladder cancer proliferation, metastasis, and angiogenesis through the PI3K/AKT pathway
Hou T, et al.
Cellular Physiology and Biochemistry, 47(6), 2250-2260 (2018)
Leupaxin is a critical adaptor protein in the adhesion zone of the osteoclast
Gupta A, et al.
Journal of Bone and Mineral Research, 18(4), 669-685 (2003)
Leupaxin, a novel coactivator of the androgen receptor, is expressed in prostate cancer and plays a role in adhesion and invasion of prostate carcinoma cells
Kaulfuss S, et al.
Molecular Endocrinology, 22(7), 1606-1621 (2008)
Structural basis for the interaction between Pyk2-FAT domain and leupaxin LD repeats
Vanarotti MS, et al.
Biochemistry, 55(9), 1332-1345 (2016)
Paxillin: a focal adhesion-associated adaptor protein
Schaller MD, et al.
Oncogene, 20(44), 6459-6472 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service