Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200004

Sigma-Aldrich

Anti-Exportin 5 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Exp5, Anti-Ran-binding protein 21, Anti-XPO5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~135 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5 μg/mL using paraformaldehyde fixed HEK-293T cells
western blot: 1-2 μg/mL using A431 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPO5(57510)
mouse ... Xpo5(72322)
rat ... Xpo5(363194)

General description

Exportin 5, also known as XPO5, Exp5 or Ran-binding protein 21, is encoded by the gene mapped to human chromosome 6p21.1. XPO5 belongs to the human karyopherin (Kap)β family. Structurally, Exp-5 has a ring-shaped closed conformation.

Specificity

Anti-Exportin 5 (N-terminal), recognizes human Exportin 5.

Application

Anti-Exportin 5 (N-terminal) antibody produced in rabbit is suitable for use in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Exportin 5 exports RNA such as viral hairpin RNAs, tRNAs and pre-microRNAs, which are likely to be its major endogenous cellular RNA cargos. Loss-of-function mutation in the gene leads to microsatellite instability.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exportin-5 mediates the nuclear export of pre-microRNAs and short hairpin RNAs
Yi R, et al.
Genes & Development, 17(24), 3011-3016 (2003)
Structural basis for selective binding of export cargoes by exportin-5
Yamazawa R, et al.
Structure, 26(10), 1393-1398 (2018)
Therapeutic targeting of nuclear export inhibition in lung cancer
Gupta A, et al.
Journal of Thoracic Disease, 12(9), 1446-1450 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service