Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200003

Sigma-Aldrich

Anti-Exportin 5 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-EXp5, Anti-Ran-binding protein 21, Anti-XPO5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~135 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5 μg/mL using paraformaldehyde fixed HEK-293T cells
western blot: 1-2 μg/mL using A431 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPO5(57510)
mouse ... Xpo5(72322)
rat ... Xpo5(363194)

General description

The exportin 5 (XPO5) gene is mapped to human chromosome 6p21.1 and belongs to the karyopherin family. Exportin 5 (also known as XPO5, Exp5, Ran-binding protein 21) is identified as a nuclear receptor for interleukin enhancer binding factor 3 (ILF3), a double-stranded RNA binding protein (dsRBP) and for several other dsRBPs.

Application

Anti-Exportin 5 (C-terminal) antibody produced in rabbit has been used in western blotting and immunofluorescence.

Biochem/physiol Actions

Exportin 5 (XPO5) plays an important role in microRNA (miRNA) cellular processing for the production of mature miRNAs. The encoded protein It transports miRNA from the nucleus to the cytoplasm in RNA-guanosine triphosphate (GTP) dependent manner. Impaired XPO5 gene expression is observed in many types of cancer. Exportin 5 can also export RNA such as viral hairpin RNAs, some tRNAs and pre-microRNAs, which are likely to be its major endogenous cellular RNA cargos.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Terminal Minihelix, a Novel RNA Motif That Directs Polymerase III Transcripts to the Cell Cytoplasm TERMINAL MINIHELIX AND RNA EXPORT
Gwizdek C, et al.
Test, 276(28), 25910-25918 (2001)
Impaired XPO5 function leads to transportation failure of microRNA precursor
Wei YQ
Science China: Life Sciences, 60(6), 677-679 (2017)
Clinical and pathological phenotype of genetic causes of focal segmental glomerulosclerosis in adults
Lepori N, et al.
Clinical kidney journal, 11(2), 179-190 (2018)
XPO5 genetic polymorphisms in cancer risk and prognosis
Patrao AS, et al.
Pharmacogenomics, 19(9), 799-808 (2018)
Toshihiko Takeiwa et al.
Genes to cells : devoted to molecular & cellular mechanisms, 20(4), 281-291 (2015-02-07)
The signal recognition particle is a ribonucleoprotein complex that is essential for the translocation of nascent proteins into the endoplasmic reticulum. It has been shown that the RNA component (SRP RNA) is exported from the nucleus by CRM1 in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service