Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200001

Sigma-Aldrich

Anti-POU1F1/PIT1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GHF-1, Anti-Pituitary-specific positive transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~32 kDa

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using GH3 cell lysates
western blot: 1-2 μg/mL using GH3 cell lysates

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POU1F1(5449)
mouse ... Pou1f1(18736)
rat ... Pou1f1(25517)

General description

POU class 1 homeobox 1 (POU1F1/ Pit1), a POU-homeodomain transcription factor is mapped on human chromosome 3p11.2. It belongs to the POU family of transcription factors.

Specificity

Anti-POU1F1/PIT1 (C-terminal) recognizes human POU1F1/PIT1.

Application

Anti-POU1F1/PIT1 (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

POU class 1 homeobox 1 (POU1F1/PIT1) plays a key role in pituitary development and hormone expression in mammals. It participates in the differentiation and proliferation of the anterior pituitary somatotrophs, lactotrophs, and thyrotropes cell lineages. Therefore, abnormalities of the Pit-1 gene results in the syndrome of combined pituitary hormone deficiency (CPHD), a disease characterized by the lack of prolactin, growth hormone, and thyroid stimulating hormone. POU1F1/PIT1 regulates its target gene expression by binding to response elements on their promoter regions and recruitment of co-regulatory proteins that alter histone acetylation and modify chromatin structures.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galia Gat-Yablonski et al.
American journal of medical genetics. Part A, 155A(9), 2242-2246 (2011-08-05)
Microdeletion syndromes include numerous syndromic phenotypes associated with intellectual disability and dysmorphic features. We report on a patient with a novel microdeletion of chromosomal region 3p11.2-p12.1 containing POU1F1, chromatin-modifying protein 2B (CHMP2B), and vestigial-like 3 (VGLL3) genes. Our patient was
B I Hendriks-Stegeman et al.
The Journal of clinical endocrinology and metabolism, 86(4), 1545-1550 (2001-04-12)
The POU homeodomain containing transcriptional activator POU1F1, formerly called Pit1 or GHF-1, is required for the embryological determination and postnatal secretory function of the GH-, PRL-, and TSH-producing cells in the anterior pituitary. Several mutations in the gene encoding POU1F1
Yingchuan Qi et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(7), 2481-2486 (2008-02-15)
Enhancers have been functionally described for >35 years, but the molecular principles underlying the integration of regulatory inputs to alternate gene enhancers used during mammalian organogenesis remain incompletely understood. Using a combination of in vivo enhancer mapping and proteomics approaches

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service