Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4100495

Sigma-Aldrich

Monoclonal Anti-MTIF3 antibody produced in mouse

clone MTIF3 -A420, culture supernatant

Synonym(s):

Anti-IF3mt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MTIF3 -A420, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 32.9 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MTIF3(219402)

General description

Mitochondrial translational initiation factor 3 (MTIF3) gene is mapped to human chromosome 13q12.2.

Immunogen

MTIF3 purified recombinant protein

Biochem/physiol Actions

Mitochondrial translational initiation factor 3 (MTIF3) initiates the synthesis of mitochondrial proteins. Altered gene function affects the expression of the mitochondrial proteins. Mutation in MTIF3 is known to cause Parkinson′s disease.

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent Findings on the Genetics of Obesity: Is there Public Health Relevance?
Richmond R C and Timpson N J
Current Nutrition Reports, 1(4), 239-248 (2012)
Genetic association analysis of 13 nuclear-encoded mitochondrial candidate genes with type II diabetes mellitus: the DAMAGE study
Reiling E, et al.
European Journal of Human Genetics, 17(8), 1056-1056 (2009)
Mitochondrial translation initiation factor 3 polymorphism and Parkinson's disease
Behrouz B, et al.
Neuroscience Letters, 486(3), 228-230 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service