Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4100331

Sigma-Aldrich

Monoclonal Anti-HARS2 antibody produced in mouse

clone HARS2-A281, culture supernatant

Synonym(s):

Anti-HARSL, Anti-HARSR, Anti-HO3, Anti-Histidyl-tRNA synthetase 2, mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

HARS2-A281, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 53.3 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... HARS2(23438)

Immunogen

HARS2 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service