Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4100257

Sigma-Aldrich

Monoclonal Anti-FADD antibody produced in mouse

clone FADD-A226, culture supernatant

Synonym(s):

AntiFas (TNFRSF6)-associated via death domain, AntiGIG3, AntiMORT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

FADD-A226, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 23.1 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FADD(8772)

Immunogen

FADD purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Yang Wang et al.
Clinical & experimental ophthalmology, 42(6), 529-538 (2013-11-15)
The aim of the study was to investigate, using a native mitomycin-C-resistant human Tenon's fibroblast cell line, the possibility that interferon-alpha and gamma could be used with Fas agonists as an alternative anti-fibrotic strategy to mitomycin-C in trabeculectomy. A clinically
Changyou Li et al.
Molecular oncology, 8(7), 1220-1230 (2014-05-13)
Little is known regarding molecular markers in head and neck squamous cell carcinoma (HNSCC) that predict responsiveness to different therapeutic regimens or predict HNSCC progression. Mutations in procaspase-8 occur in 9% of HNSCC primary tumors, but the functional consequences of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service