Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4100228

Sigma-Aldrich

Monoclonal Anti-PTPRE antibody produced in mouse

clone PTPRE-B186, culture supernatant

Synonym(s):

AntiHPTPE, AntiPTPE, AntiProtein tyrosine phosphatase, receptor type, E, AntiR-PTP-EPSILON

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

PTPRE-B186, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 80.6 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

Gene Information

human ... PTPRE(5791)

General description

PTPRE (Protein tyrosine phosphatase, receptor type, E) is a transmembrane protein consisting of a short extracellular domain and two tandemly repeated intracellular PTPase domains. It is located on chromosome 10q26 in humans. It is expressed in testis, brain, lung and lymph node. Its expression also has been observed during neuronal differentiation and in murine mammary tumour cells. It has an isoform consisting of an N-terminal catalytic, active phosphatase domain 1 (PD1).

Immunogen

PTPRE purified recombinant protein

Application

Monoclonal Anti-PTPRE antibody produced in mouse is suitable for western blot assay.

Biochem/physiol Actions

PTPRE (Protein tyrosine phosphatase, receptor type, E) is associated with various signalling pathways including down regulation of insulin receptor signalling and Janus kinase (Jak)-STAT inhibition signalling pathway. It also plays an important role in the regulation of ERK1/2 pathway by emerging as a phosphatase through its catalytic activity. Study shows that it may play an accessory role in tumour development.

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of the human protein tyrosine phosphatase epsilon (PTPRE) gene to chromosome 10q26 by fluorescence in situ hybridization.
A M van den Maagdenberg et al.
Genomics, 30(1), 128-129 (1995-11-01)
T Wabakken et al.
Scandinavian journal of immunology, 56(3), 276-285 (2002-08-24)
This report describes the identification and characterization of a novel cytoplasmic isoform of human protein tyrosine phosphatase epsilon (PTPepsilon). The novel isoform, denoted cyt-PTPepsilonPD1, displays only the N-terminal catalytic, active phosphatase domain 1 (PD1) which is common in all known
T Wabakken et al.
Scandinavian journal of immunology, 56(2), 195-203 (2002-07-18)
The expression of protein tyrosine phosphatase epsilon (PTPepsilon) was studied in human tissues and blood cells. High mRNA expression was observed in peripheral blood leucocytes, particularly in monocytes and granulocytes which revealed at least four distinct transcripts. In lymphocytes, PTPepsilon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service