SAB4100218
Monoclonal Anti-PBLD antibody produced in mouse
clone PBLD-A170, culture supernatant
Synonym(s):
AntiMAWBP, AntiMAWD binding protein, AntiMAWDBP, AntiOTTHUMP00000019698, AntiPhenazine biosynthesis-like protein domain containing
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
PBLD-A170, monoclonal
form
aqueous solution
mol wt
antigen predicted mol wt 31.6 kDa
species reactivity
human
technique(s)
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... PBLD(64081)
Immunogen
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service