Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4100211

Sigma-Aldrich

Monoclonal Anti-VAT1L antibody produced in mouse

clone VAT1L-A56, culture supernatant

Synonym(s):

AntiVesicle amine transport protein 1 homolog (T. californica)-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

VAT1L-A56, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 45.7 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... VAT1L(57687)

Immunogen

VAT1L purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service