Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4100165

Sigma-Aldrich

Monoclonal Anti-MAPK7 antibody produced in mouse

clone MAPK7-B159, culture supernatant

Synonym(s):

Anti-BMK1, Anti-ERK4, Anti-ERK5, Anti-Extracellular-signal-regulated kinase 5, Anti-Mitogen-activated protein kinase 7, Anti-OTTHUMP00000065906big MAP kinase 1, Anti-PRKM7, Anti-big MAP kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MAPK7-B159, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 88.2 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MAPK7(5598)

Immunogen

MAPK7 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufeng Zuo et al.
Journal of cellular biochemistry, 116(1), 124-132 (2014-08-28)
Members of Rho family GTPases including Cdc42 are known to play pivotal roles in cell migration. Cell migration is also known to be regulated by many protein kinases. Kinetworks KPSS 11.0 phospho-site screening of Cdc42-silenced Hs578T breast cancer cells revealed
Nayara I Viana et al.
The International journal of biological markers, 29(3), e246-e252 (2014-01-30)
The aim of this study was to analyze the roles of miR-143 and miR-145, as well as the gene and protein expression of their targets (KRAS, ERK5, MAP3K3, and MAP4K4) in the pathogenesis of benign prostatic hyperplasia (BPH). We analyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service