SAB4100059
Monoclonal Anti-MIOX antibody produced in mouse
clone MIOX-C69, culture supernatant
Synonym(s):
Anti-ALDRL6, Anti-Aldehyde reductase (aldose reductase) like 6, Anti-Kidney-specific protein 32, Anti-Myo-inositol oxygenase
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
MIOX-C69, monoclonal
form
aqueous solution
mol wt
antigen predicted mol wt 32.8 kDa
species reactivity
human
technique(s)
western blot: suitable
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... MIOX(55586)
Looking for similar products? Visit Product Comparison Guide
Immunogen
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service