Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3701465

Sigma-Aldrich

Monoclonal Anti-TIMP4 antibody produced in mouse

clone 9:4-7, purified immunoglobulin

Synonym(s):

Anti-Death up-regulated gene protein antibody, Anti-Metalloproteinase inhibitor 4, Anti-TIMP 4, Anti-TIMP-4, Anti-TIMP4 antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9:4-7, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TIMP4(7079)

Specificity

Anti-TIMP4 was purified from clarified mouse ascetic fluid by Protein A chromatography followed by extensive dialysis against the buffer stated above. TIMP4 antibody is specific for human TIMP4 protein. This antibody reacts with human TIMP4 in tissue sections of formalin-fixed, paraffin-embedded In ELISAs and Native-PAGE, this antibody does not cross-react with recombinant human TIMP-1, -2 or -3. A BLAST analysis was used to suggest cross-reactivity with TIMP4 from human and mouse sources based on the immunizing sequence. Cross-reactivity with TIMP-4 from other sources has not been determined.

Immunogen

Full length human TIMP4 protein followed by hybridoma development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% (w/v) Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service