Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3701302

Sigma-Aldrich

Anti-Human IgG (H+L), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in donkey

affinity isolated antibody, lyophilized powder

Synonym(s):

FITC

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$290.00

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
500 μL
$290.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

donkey

Quality Level

conjugate

fluorescein conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

direct immunofluorescence: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Fluorescein, Anti-Donkey Serum, Human IgG and Human Serum.  No reaction was observed against Anti-Pepsin, Anti-Donkey IgG F(c) or Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Mouse, Rabbit, Rat and Sheep Serum Proteins.

Immunogen

Human IgG whole molecule

Physical properties

Antibody format: IgG F(ab′)2

Physical form

Supplied in 0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Reconstitution

Reconstitute with 500 μ;L deionized water (or equivalent).

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service