Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3701204

Sigma-Aldrich

Anti-Mouse IgM (μ-chain specific), F(ab′)2 fragment antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Immunoglobulin M (IgM) is a member of immunoglobulin family. It exists as a pentamer in the serum and has a high molecular weight. Monomeric IgM has a molecular weight of 190kDa. The pentameric IgM is characterized with ten μ heavy chains, ten light chains and a single J-chain which is linked by disulphide bonds. The μ chain consists of five domains- VH, Cμ1, Cμ2, Cμ3 and Cμ4. Limited digestion using papain cleaves the antibody into three fragments, two of which are identical and contain the antigen-binding activity. They are known as fragment antigen binding (Fab) fragments. The third fragment does not possess antigen-binding activity and is known as fragment crystallizable (Fc). It interacts with cells and effector molecules. The heavy chains of IgM contain an extra C domain that replaces the hinge region. IgM is the first antibody to be produced during an immune response and is the major isotype secreted in T-cell independent immune responses.

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgM coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Goat Serum, Mouse IgM and Mouse Serum. No reaction was observed against Anti-Pepsin or Anti-Goat IgG F(c). Specificity was confirmed by ELISA at less than 1% cross-reactivity against other Mouse heavy or light chain isotypes.

Immunogen

Mouse IgM mu heavy chain

Physical properties

Antibody format: IgG F(ab′)2

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum Immunoglobulin M Concentration Is Positively Related to Metabolic Syndrome in an Adult Population: Tianjin Chronic Low-Grade Systemic Inflammation and Health (TCLSIH) Cohort Study
Kun Song
PLoS ONE, 9(2), e88701-e88701 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service