Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3700991

Sigma-Aldrich

Anti-Mouse IgG F(ab′)2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$219.00

$219.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
2 MG
$219.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$219.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

2.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Goat Serum, Mouse IgG, Mouse IgG F(ab′)2 and Mouse Serum. No reaction was observed against Mouse IgG F(c).

Immunogen

Mouse IgG F(ab′)2 fragment

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service