SAB3700905
Anti-Rabbit IgG (H+L), highly cross adsorbed-Fluorescein antibody produced in rat
affinity isolated antibody, lyophilized powder
Synonym(s):
FITC
Select a Size
$239.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$239.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
rat
conjugate
fluorescein conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
rabbit
technique(s)
direct immunofluorescence: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
shipped in
wet ice
Storage temp.
2-8°C
target post-translational modification
unmodified
Related Categories
Specificity
Immunogen
Physical properties
Physical form
Reconstitution
Not finding the right product?
Try our Product Selector Tool.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2
supp_hazards
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service