Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3700840

Sigma-Aldrich

Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$286.00

$286.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
500 μG
$286.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$286.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Rabbit IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Goat Serum, Rabbit IgG, Rabbit IgG F(ab′)2 and Rabbit Serum. No reaction was observed against Anti-Pepsin, Anti-Goat IgG F(c), Rabbit IgG F(c) or Bovine, Horse, Human, Mouse, Rat and Sheep Serum Proteins.

Immunogen

Rabbit IgG F(ab′)2 fragment

Physical properties

Antibody format: IgG F(ab′)2

Physical form

Supplied in 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianwen Zeng et al.
European journal of pharmacology, 762, 82-88 (2015-05-26)
The aim of the present study was to investigate the expression and distribution of membrane receptors after bladder outlet obstruction (BOO). Partial bladder outlet obstruction (BOO) was induced in female rats and bladders were harvested after either 10 days or
Liang Sun et al.
Molecular medicine reports, 18(4), 3818-3824 (2018-08-15)
The aim of the present study was to investigate the effects of Toll‑like receptor (TLR) 9 and CpG oligodeoxynucleotide (CpG‑ODN)1826 on sodium taurocholate‑induced acute pancreatitis (AP) rats at different time points. Pathological examination indicated that the severity of pancreatic tissue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service