This product was prepared from monospecific antiserum by immunoaffinity chromatography using Sheep IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Rabbit Serum, Sheep IgG and Sheep Serum. No reaction was observed against Anti-Pepsin or Anti-Rabbit IgG F(c).
Immunogen
Sheep IgG whole molecule
Physical properties
Antibody format: IgG F(ab′)2
Physical form
Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Reconstitution
Reconstitute with 500 μ;L deionized water (or equivalent).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.