This product was prepared from monospecific antiserum by immunoaffinity chromatography using Sheep IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Rabbit Serum, Sheep IgG, Sheep IgG F(ab′)2 and Sheep Serum. No reaction was observed against Sheep IgG F(c). This antibody was tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with Human Serum Proteins, but antibodies may cross-react with immunoglobulins from other species.
Immunogen
Sheep IgG F(ab′)2 fragment
Physical properties
Antibody format: IgG
Physical form
Supplied in 0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.