Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3700631

Sigma-Aldrich

Anti-Rat IgG (H+L), F(ab′)2 fragment-Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ALP, Alk Phos

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$290.00

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
500 μG
$290.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Rat IgG coupled to agarose beads followed by pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Alkaline Phosphatase, Anti-Rabbit Serum, Rat IgG and Rat Serum. No reaction was observed against Anti-Pepsin or Anti-Rabbit IgG F(c).

Immunogen

Rat IgG whole molecule

Physical properties

Antibody format: IgG F(ab′)2

Physical form

Supplied in 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service