direct immunofluorescence: suitable indirect ELISA: suitable indirect immunofluorescence: suitable
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Specificity
This product was prepared from monospecific antiserum by immunoaffinity chromatography using Golden Syrian Hamster IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, papain digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Fluorescein and Anti-Goat Serum. No reaction was observed against Anti-Papain or Anti-Goat IgG F(c).
Immunogen
Golden Syrian Hamster IgG whole molecule
Physical properties
Antibody format: IgG Fab
Physical form
Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Reconstitution
Reconstitute with 1.0 mL deionized water (or equivalent).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.