immunohistochemistry: suitable indirect ELISA: suitable western blot: suitable
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Specificity
This product was prepared from monospecific antiserum by immunoaffinity chromatography using Golden Syrian Hamster IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Rabbit Serum, Golden Syrian Hamster IgG, Golden Syrian Hamster IgG and Golden Syrian Hamster Serum. No reaction was observed against Anti-Pepsin, Anti-Rabbit IgG F(c). Limited reactivity will occur with Armenian Hamster IgG.
Immunogen
Golden Syrian Hamster IgG whole molecule
Physical properties
Antibody format: IgG F(ab′)2
Physical form
Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.