Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3501094

Sigma-Aldrich

Anti-SIN NOMBRE VIRUS GLYCOPROTEIN 2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$398.00

$398.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$398.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$398.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

virus

concentration

1 mg/mL

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Sin Nombre virus ... GPC(2654026)

General description

Sin Nombre virus (SNV) is an enveloped virus with a tripartite negative-sense RNA genome. This viral genome codes for a nucleocapsid N protein, the glycoprotein precursor GPC and the RNA polymerase L protein. SNV belongs to the Bunyaviridae family. SNV virions are usually round in shape with a mean diameter of 112 nm. These virions have filamentous nucleocapsids.

Immunogen

Antibody was raised against a 16 amino acid peptide near the center of the Sin Nombre virus glycoprotein.

Biochem/physiol Actions

Sin Nombre virus (SNV) is a novel representative of the New World hantaviruses and the major cause of hantavirus pulmonary syndrome (HPS).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501094.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrastructural characteristics of Sin Nombre virus, causative agent of hantavirus pulmonary syndrome.
Goldsmith C S, et al.
Archives of Virology, 140(12), 2107-2122 (1995)
Sin Nombre virus glycoprotein trafficking.
Spiropoulou C F, et al.
Virology, 308(1), 48-63 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service