Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3501052

Sigma-Aldrich

Anti-AIMP2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 35 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AIMP2(7965)

General description

Aminoacyl tRNA synthetase complex interacting multifunctional protein 2 (AIMP2) is localized to the cytosol. It is part of the multi-tRNA synthetase complex. The AIMP2 gene is located on human chromosome 7.

Immunogen

Antibody was raised against a 16 amino acid peptide near the center of human AIMP2.

Biochem/physiol Actions

Aminoacyl tRNA synthetase complex interacting multifunctional protein 2 (AIMP2) is a scaffold protein which functions as a tumor suppressor. It stimulates the ubiquitin-mediated degradation of FUSE-binding protein (FUBP1), which is a transcription activator. AIMP2 activates tumor suppressor p53 and stimulates apoptosis. The assembly of macromolecular tRNA synthetase complex depends on AIMP2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501052.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer-associated splicing variant of tumor suppressor AIMP2/p38: pathological implication in tumorigenesis.
Choi JW
PLoS Genetics, 7(3), e1001351-e1001351 (2011)
Dae Gyu Kim et al.
Cancer research, 76(11), 3422-3436 (2016-05-20)
AIMP2/p38 is a multifunctional tumor suppressor that normally resides in the cytosol as a scaffold protein of the multi-tRNA synthetase complex (MSC). One of the tumor-suppressive functions of AIMP2 is to facilitate ubiquitin-mediated degradation of FUSE-binding protein (FBP, FUBP1), a
You-Sub Won et al.
Journal of biotechnology, 158(1-2), 44-49 (2012-01-31)
AIMP2/p38 is a scaffolding protein critical for the assembly of the macromolecular tRNA synthetase complex. Moreover, AIMP2 harbors anti-proliferative activity and promotes cell death as a proapototic factor. A splicing variant of AIMP2 lacking exon 2 (AIMP2-DX2) is specifically generated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service