Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500559

Sigma-Aldrich

Anti-KPNA1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IPOA5, Anti-Importin alpha 5, Anti-Karyopherin alpha 1 Nucleoprotein interactor 1, Anti-RAG cohort protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$405.00

$405.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$405.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$405.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KPNA1(3836)

Immunogen

KPNA1 antibody was raised against a 15 amino acid peptide near the amino terminus of human KPNA1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500559.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service