Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3500461

Sigma-Aldrich

Anti-TL1A antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-TL1A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 28 kDa

species reactivity

mouse, rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFSF15(9966)

General description

Tumor necrosis factor ligand superfamily member 15 (TNFSF15) or TNF-like ligand 1A (TL1A) is a 174 amino acid, type II transmembrane protein. The molecular mass of the protein is 13kDa and it belongs to the tumor necrosis factor (TNF) family. TNFSF15 has a carboxyl terminal which is present on the exterior of the cell surface, a transmembrane domain and a short cytoplasmic tail. The gene encoding it is located on human chromosome 9q32.

Immunogen

TL1A antibody was raised against a peptide corresponding to 14 amino acids near the N-terminus of human TL1A.

Biochem/physiol Actions

Tumor necrosis factor ligand superfamily member 15 (TNFSF15)/TNF-like ligand 1A (TL1A) functions as an angiogenesis inhibitor by inhibiting the endothelial cell proliferation. It may play a role in angiogenesis-based cancer therapy. TNFSF15 activates the transcription factor κ B (NF-κB). It induces the degradation of its inhibitor (IκBα) and also aids the nuclear translocation of the p65 subunit of NF-κ B. c-Jun N-terminal kinase activation is initiated by TNFSF15 and it also inhibits the growth of various human tumor cell lines. Single nucleotide polymorphisms in the gene encoding this protein are associated with surgical diverticulitis and susceptibility to ulcerative colitis and Crohn′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500461.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VEGI, a new member of the TNF family activates nuclear factor-kappa B and c-Jun N-terminal kinase and modulates cell growth.
Oncogene (1999)
VEGI, a novel cytokine of the tumor necrosis factor family, is an angiogenesis inhibitor that suppresses the growth of colon carcinomas in vivo.
Zhai Y
Faseb Journal (1999)
The TNFSF15 gene single nucleotide polymorphism rs7848647 is associated with surgical diverticulitis.
Connelly TM
Annals of Surgery (2014)
Associations between TNFSF15 polymorphisms and susceptibility to ulcerative colitis and Crohn's disease: A meta-analysis.
Zhang J
Autoimmunity (2014)
Shunsuke Kumanishi et al.
International journal of oncology, 55(1), 167-178 (2019-06-11)
Vascular endothelial growth inhibitor (VEGI; also referred to as TNFSF15 or TL1A) is involved in the modulation of vascular homeostasis. VEGI is known to operate via two receptors: Death receptor‑3 (DR3) and decoy receptor‑3 (DcR3). DR3, which is thus far

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service