Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500455

Sigma-Aldrich

Anti-Rim3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NIM3, Anti-RIM3 gamma, Anti-RIMS3, Anti-Rab3-interacting molecule 3, Anti-Regulating synaptic membrane exocytosis 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$545.00

$545.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$545.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$545.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIMS3(9783)

Immunogen

Rim3 antibody was raised against a 17 amino acid peptide from near the amino terminus of human Rim3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500455.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanyan Zhang et al.
Journal of tissue engineering and regenerative medicine, 13(2), 244-252 (2018-12-18)
The contents of platelet α-granules arrive via a number of pathways; some are synthesized by megakaryocytes (MKs), for example, von Willebrand factor (VWF), whereas others are endocytosed from plasma, for example, fibrinogen (Fgn) and factor V (FV). Currently, almost all

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service