Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3500105

Sigma-Aldrich

Anti-RWDD3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution, ab2

Synonym(s):

Anti-RSUME, Anti-RWD domain containing 3, Anti-RWD-containing sumoylation enhancer

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$493.00

$493.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$493.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$493.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RWDD3(25950)

Immunogen

RWDD3 antibody was raised against a 17 amino acid peptide from near the amino terminus of human RWDD3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

RWDD3 (RSUME), a small RWD-containing protein, has a central role in sumoylation by enhancing SUMO conjugation in the regulatory network of immune–inflammatory signals. RWDD3 increases Ikbeta sumoylation and stability. In addition, RWDD3 inhibits TNF-alpha-induced kB-LUC (Luciferase) reporter activity, showing the functional consequence of Ikbeta increased stability. RSUME-enhanced sumoylation of Ikbeta leads to the inhibition of NF-kbeta activity on two well-known inflammatory genes, IL-8 and cyclooxygenase-2 (Cox-2) and therefore may also favor anti-inflammatory pathways. Expression of RWDD3 was induced under hypoxic conditions and it has a potential role during vascularization. Both BMP-4 and RWDD3 may be interesting targets for inhibiting steps involved in pituitary tumorigenesis.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500105.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service