SAB3500092
Anti-Seasonal H1N1 Nucleocapsid Protein antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Common flu NP, Anti-NP
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
viral
technique(s)
indirect ELISA: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
General description
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus (S-OIV) was identified in specimens obtained from patients in Mexico and the United States. The influenza A virus polymerase transcribes and replicates eight virion RNA (vRNA) segments. The nucleocapsid protein (NP) is produced among these segments. It is thought to control whether mRNA or cRNA is produced. NP has multiple functions during the virus life cycle. It possesses regions that are highly conserved among influenza A, B and C viruses. It is studied that NP mutations affected the efficient incorporation of multiple viral-RNA (vRNA) segments into progeny virions. This indicates that the conserved amino acids in NP might be critical for the assembly and incorporation of sets of eight vRNA segments. This antibody is specific for the seasonal H1N1 influenza NP and will not recognize the corresponding NP sequence from the swine-origin H1N1 influenza (A/California/14/2009 (H1N1)).
Immunogen
NP antibody was raised against a synthetic peptide from the Seasonal H1N1 NP protein.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Target description
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus (S-OIV) was identified in specimens obtained from patients in Mexico and the United States. The influenza A virus polymerase transcribes and replicates eight virion RNA (vRNA) segments, among which the nucleocapsid protein (NP), thought to control whether mRNA or cRNA is produced. The nucleoprotein (NP), which has multiple functions during the virus life cycle, possesses regions that are highly conserved among influenza A, B, and C viruses. It was recently found several NP mutations that affected the efficient incorporation of multiple viral-RNA (vRNA) segments into progeny virions even though a single vRNA segment was incorporated efficiently. This indicates that the respective conserved amino acids in NP may be critical for the assembly and/or incorporation of sets of eight vRNA segments. This antibody is specific for the seasonal H1N1 influenza NP and will not recognize the corresponding NP sequence from the swine-origin H1N1 influenza (A/California/14/2009 (H1N1)).
Linkage
The action of this antibody can be blocked using blocking peptide SBP3500092.
Physical form
Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Interaction of the influenza a virus nucleocapsid protein with the viral RNA polymerase potentiates unprimed viral RNA replication.
Newcomb L L, et al.
Journal of Virology, 83(1), 29-36 (2009)
Mutational analysis of conserved amino acids in the influenza A virus nucleoprotein.
Li Z, et al.
Journal of Virology, 83(9), 4153-4162 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service