Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500081

Sigma-Aldrich

Anti-SCAMP4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Secretory carrier membrane protein 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$370.00

$370.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$370.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCAMP4(113178)

Immunogen

SCAMP4 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human SCAMP4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Secretory carrier membrane proteins (SCAMPs) are integral membrane proteins seen in secretory and endocytic carriers that are thought to play roles in membrane trafficking. One member of this family, SCAMP4, lacks the putative calcium binding and leucine zipper structures as well as the protein-protein interacting NPF domains common to most SCAMP family members. In rats, SCAMP4 mRNA is repressed by progesterone in brain regions associated with female sexual behavior. At least three isoforms of SCAMP4 are known to exist. This SCAMP4 antibody is predicted to not cross-react with any other member of the secretory carrier membrane protein family.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500081.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service