SAB3500077
Anti-MAK10 antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution, ab1
Synonym(s):
Anti-EGAP, Anti-Embryonic growth-associated protein homolog, Anti-amino-acid N-acetyltransferase subunit
Select a Size
$370.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$370.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human, mouse, rat
technique(s)
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MAK10(60560)
Immunogen
Features and Benefits
Target description
Linkage
Physical form
Not finding the right product?
Try our Product Selector Tool.
related product
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service