Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500076

Sigma-Aldrich

Anti-MPYS antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERIS, Anti-MITA, Anti-STING, Anti-TMEM173, Anti-Transmembrane protein 173

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 31-42 kDa

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

TMEM173 (transmembrane protein 173) gene is mapped to human chromosome 5q31.2. The encoded protein localizes in the endoplasmic reticulum.

Immunogen

MPYS antibody was raised against a 17 amino acid peptide near the carboxy terminus of human MPYS.

Biochem/physiol Actions

The innate immune response against the viral and bacterial infections is mostly regulated by TMEM173 (transmembrane protein 173), also known as stimulator of interferon genes (STING). It is considered as an important adaptor protein for cytosolic DNA sensing pathways. Downregulation of the gene was observed in tumor tissues. TMEM173 acts as a pattern recognition receptor and identifies pathogen-associated molecular patterns. It also mediates signal transduction pathways. TMEM173 is significantly associated with immunity and inflammation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

MPYS is a recently identified plasma membrane tetraspanner that is associated with major histocompatibility complex class II (MHC-II) and mediates its transduction of apoptotic signals. It has also been found to be associated with VISA, a mitochondrial protein that acts as an adaptor in virus-triggered signaling. MPYS also interacts with IRF3 and recruits the kinase TBK1 to the VISA-associated complex, acting as a critical mediator of virus-triggered IRF3 activation and interferon (IFN) expression. It is thought that the binding of nucleic acid to the innate immune protein RIG-I causes complex formation between RIG-I, VISA, and MPYS. This complex then recruits TBK1 to phosphorylate IRF3 which then directly activates IFN transcription. At least three isoforms of MPYS are known to exist.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500076.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HSV-1 ICP27 targets the TBK1-activated STING signalsome to inhibit virus-induced type I IFN expression.
Christensen MH
The Embo Journal (2016)
DNA exonuclease Trex1 regulates radiotherapy-induced tumour immunogenicity.
Vanpouille-Box C
Nature Communications (2017)
Retroviral insertional mutagenesis identifies the del(5q) genes, CXXC5, TIFAB and ETF1, as well as the Wnt pathway, as potential targets in del(5q) myeloid neoplasms.
Stoddart A
Haematologica (2016)
STING-IRF3 Triggers Endothelial Inflammation in Response to Free Fatty Acid-Induced Mitochondrial Damage in Diet-Induced Obesity.
Mao Y
Arteriosclerosis, Thrombosis, and Vascular Biology (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service