Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3500050

Sigma-Aldrich

Anti-UCMA antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Upper zone of growth plate and cartilage matrix associated

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$370.00

$370.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$370.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 15 kDa

species reactivity

human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCMA(221044)

Immunogen

UCMA antibody was raised against a 16 amino acid peptide near the carboxy terminus of human UCMA.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

UCMA is a secreted cartilage-specific protein that was discovered in a screen for differentially expressed genes in retinoic acid-treated mouse chondrocytes. It was also identified in a human chondrocyte EST screen for candidate genes of skeletal dysplasias. UCMA expression is thought to parallel that of collagen II with its expression decreasing with maturation chrondrocytes mature. UCMA is processed by a furin-like protease into two fragments, an amino-terminal fragment and a carboxy-terminal fragment (UCMA-C). Application of recombinant UCMA-C to primary osteoblasts, mesenchymal stem cells, and MC3T3-E1 pre-osteoblasts interferes with their osteogenic differentiation, but does not affect expression of chondrocyte-specific genes or chondrocyte proliferation, suggesting that UCMA may be involved in the negative control of osteogenic differentiation of osteochondrogenic precursor cells. At least two isoforms of UCMA are known to exist.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500050.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service