Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900851

Sigma-Aldrich

Anti-SYPL2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MG29, Mitsugumin 29, Mitsugumin-29, SYPL2, Synaptophysin-like 2, Synaptophysin-like protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

western blot: 1-2 μg/mL (Recombinant Protein)
western blot: 5 μg/mL (Cell Lysate)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SYPL2(284612)

Immunogen

SYPL1 antibody was raised against a 16 amino acid peptide from near the amino terminus of human SYPL1.
SYPL2 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human SYPL.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service