Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900206

Sigma-Aldrich

Anti-BACE1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

APP beta-secretase, asp2, aspartylprotease 2, b-Secretase, bace

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, bovine, hamster, guinea pig, dog, pig, mouse, horse, monkey, rat, rabbit

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BACE1(23621)

Immunogen

Synthetic 15 amino acid peptide from internal region of human BACE1. Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Opossum, Guinea pig (100%); Platypus (87%); Xenopus, Pufferfish, Zebrafish, Stickleback (80%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service