Solute carrier family 7, membrane 11 (SLC7A11), a constituent of the cysteine-glutamate transporter, is located on human chromosome 4q28.31. It belongs to the system xc- light chain family. SLC7A11 gene codes for the cystine-glutamate antiporter Xct.
Immunogen
Synthetic 19 amino acid peptide from N-terminus of human SLC7A11. Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon (100%); Monkey, Marmoset (95%); Horse (89%); Rat, Dog, Hamster (84%).
Application
Anti-SLC7A11 antibody has been used in western blot analysis.
Biochem/physiol Actions
Solute carrier family 7, membrane 11 (SLC7A11) or xCT participates in glutathione homeostasis, that helps to guard the cells against oxidative stress. It is involved in the pathogenesis of malignant tumors. SLC7A11 is associated with triple negative breast cancer and glioblastoma (GBM).
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate-buffered saline containing less than 0.1% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
We aimed to detect the functions of miR-375/SLC7A11 axis on oral squamous cell carcinoma (OSCC) cell proliferation and invasion. Expression levels of miR-375 and SLC7A11 in OSCC tissues and cells were measured with RT-qPCR and western blot. Targeting site was
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.