Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900061

Sigma-Aldrich

Anti-CAV1 (AB1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

caveolin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, monkey, dog, rat, horse, bovine, sheep, human, rabbit, mouse, guinea pig

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CAV1(857)

General description

Caveolin 1 (CAV1) is a caveolae coat protein belonging to the caveolin family. It is expressed on fibroblasts, adipocytes and endotheliocytes. The protein possesses a highly conserved caveolin scaffolding domain (CSD). The gene encoding CAV1 is localized on human chromosome 7q31.1–31.2 and consists of 3 exons.

Immunogen

Synthetic 15 amino acid peptide from N-terminus of human CAV1 / Caveolin. Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Baboon, Monkey, Galago, Marmoset, Mouse, Rat, Ferret, Sheep, Bovine, Dog, Cat, Bat, Horse, Rabbit, Pig, Guinea pig (100%); Elephant, Armadillo (93%).

Biochem/physiol Actions

Caveolin 1 (CAV1) is involved in cell proliferation, migration, differentiation and apoptosis. Its abnormal expression has been linked to the development of tumors. CAV1 has also been shown to be associated with various cancers like gastric, breast and prostate cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The phospho-caveolin-1 scaffolding domain dampens force fluctuations in focal adhesions and promotes cancer cell migration.
Meng F
Molecular Biology of the Cell, 28(16), 2190-2201 (2017)
The influence of caveolin-1 gene polymorphisms on hepatitis B virus-related hepatocellular carcinoma susceptibility in Chinese Han population: A case-control study.
Zhang J
Medicine, 96(42) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service