Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2900026

Sigma-Aldrich

Anti-GPR89A antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

GPHR, Golgi pH regulator, GPR89, cgi-13, g protein-coupled receptor sh120, sh120, sh120 g protein-coupled receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, horse, pig, bovine, monkey, hamster, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GPR89A(653519)

General description

G protein-coupled receptor 89A (GPR89A) has been shown to be overexpressed in breast cancer. The gene encoding this protein is localized on human chromosome 1q21.1.

Immunogen

Synthetic 17 amino acid peptide from cytoplasmic loop of human GPR89A. Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Hamster, Horse, Pig, Opossum (100%); Bat, Panda, Chicken, Platypus, Xenopus, Pufferfish, Zebrafish, Stickleback (94%); Dog, Salmon (88%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigating the genetic basis of fever-associated syndromic epilepsies using copy number variation analysis
Corinna Hartmann
Epilepsia, 56(3) (2015)
UK Breast Cancer Research Symposium 2016: Submitted
Abstracts
Breast Cancer Research and Treatment, 177-197 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service