Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2702536

Sigma-Aldrich

Anti-beta Lactoglobulin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

LGB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

bovine

concentration

1mg/mL

technique(s)

ELISA: suitable
FUNC: suitable
western blot: 1:5000-1:20000

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... PAEP(280838)

Immunogen

Rabbits were immunized with bovine beta lactoglobulin. After multiple immunization in Freund′s adjuvant the rabbits were bled, serum collected and the antibodies isolated by immunoaffinity chromatography

Application

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, ELISA. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Primary component of whey, it binds retinol and is probably involved in the transport of that molecule.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Purified by antigen-affinity chromatography

Physical form

1XPBS. 0.1% Sodium Azide was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ricardo N Pereira et al.
Food & function, 11(5), 4002-4013 (2020-04-23)
β-Lactoglobulin (β-LG) encompasses important biological functions, but it is also a major milk allergen, being responsible for the high incidence of cow's milk allergy in childhood. Despite numerous studies addressing the effect of thermal processing on the immunoreactivity of β-LG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service