Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2702462

Sigma-Aldrich

Anti-Alcama antibody produced in rabbit

affinity isolated antibody

Synonym(s):

DM-GRASP, alcam, cd166

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61kDa

species reactivity

zebrafish

concentration

1mg/mL

technique(s)

immunohistochemistry: suitable

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... alcama(30194)

General description

Alcama is a member of immunoglobulin (Ig) superfamily and is expressed in the pharyngeal endoderm.[1]

Immunogen

Recombinant protein encompassing a sequence within the center region of zebrafish Alcama.

Application

Suggested starting dilutions are as follows: IHC-Wm: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Alcama has a role in neurogenesis and mediates axon growth and guidance.[1]
Cell adhesion molecule. Involved in neurite extension by neurons via heterophilic and homophilic interactions.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Purified by antigen-affinity chromatography

Physical form

1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alcama mediates Edn1 signaling during zebrafish cartilage morphogenesis.
Choudhry P, et al.
Developmental Biology, 349(2), 483-493 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service